Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-T-23 | 171674-76-3 | 96.1% | 480.43 g/mol | C23H20N4O8 | 5MG
SDP

Catalog No. 5000238839
Encompass_Preferred

Bis-T-23 (AG1717) is a research small molecule reported as an HIV-1 integrase inhibitor that also promotes actin-dependent dynamin oligomerization. Supplied as a purified powder with supporting documentation, it is used in biochemical and cell biology studies to investigate integrase function and cytoskeletal dynamics.

  • Purity: 96.1%.
  • Molecular weight: 480.43 g/mol.
  • Chemical formula: C23H20N4O8.
  • CAS number: 171674-76-3.
  • Recommended storage: powder -20°C up to 3 years; in solvent -80°C up to 6 months.
  • Available vial sizes: 1 mg, 5 mg, 10 mg, 25 mg; larger sizes by quote.
  • Documentation available: data sheet, certificate of analysis, safety data sheet.

Catalog No. 50-002-38839 Supplier Medchemexpress LLC Supplier No. HY1235725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-T-23 (AG1717) is a research small molecule reported as an HIV-1 integrase inhibitor that also promotes actin-dependent dynamin oligomerization. Supplied as a purified powder with supporting documentation, it is used in biochemical and cell biology studies to investigate integrase function and cytoskeletal dynamics.

  • Purity: 96.1%.
  • Molecular weight: 480.43 g/mol.
  • Chemical formula: C23H20N4O8.
  • CAS number: 171674-76-3.
  • Recommended storage: powder -20°C up to 3 years; in solvent -80°C up to 6 months.
  • Available vial sizes: 1 mg, 5 mg, 10 mg, 25 mg; larger sizes by quote.
  • Documentation available: data sheet, certificate of analysis, safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.