Learn More
TARGETMOL CHEMICALS INC BIS-T-23 5MG
SDP
Catalog No. 502580414
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Bis-T-23 is a promoter of actin-dependent dynamin oligomerization an HIV-I integrase inhibitor and a Telstar derivative. Bis-T-23 promotes actin-dependent dynamin oligomerization. Bis-T-23 can be used in studies of HIV and chronic kidney disease (CKD). purity: 98%
Catalog No. 50-258-0414 Supplier TARGETMOL CHEMICALS INC Supplier No. T304795MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Bis-T-23 is a promoter of actin-dependent dynamin oligomerization an HIV-I integrase inhibitor and a Telstar derivative. Bis-T-23 promotes actin-dependent dynamin oligomerization. Bis-T-23 can be used in studies of HIV and chronic kidney disease (CKD). purity: 98%