Learn More
Medchemexpress LLC Bis-T-23 (AG1717) | 171674-76-3 | 96.1% | 480.43 g/mol | C23H20N4O8 | 1MG
SDP
Catalog No. 5000236692
Encompass_Preferred
Bis-T-23 (AG1717) is a tyrphostin-derived small-molecule research compound described as an HIV-1 integrase inhibitor that promotes actin-dependent dynamin oligomerization. Supplied for laboratory research use only, it is used in studies of HIV infection and chronic kidney disease, and is provided in milligram-scale quantities for biochemical and cellular assays.
  • Promotes actin-dependent dynamin oligomerization.
  • Reported HIV-1 integrase inhibitory activity.
  • Powder stable at -20°C for up to 3 years.
  • In-solution stability: -80°C for 6 months, -20°C for 1 month.
  • Available in small-mass pack sizes suitable for research assays.
  • Molecular formula C23H20N4O8; molecular weight 480.43 g/mol.
Catalog No. 50-002-36692 Supplier Medchemexpress LLC Supplier No. HY1235721MG
Bis-T-23 (AG1717) is a tyrphostin-derived small-molecule research compound described as an HIV-1 integrase inhibitor that promotes actin-dependent dynamin oligomerization. Supplied for laboratory research use only, it is used in studies of HIV infection and chronic kidney disease, and is provided in milligram-scale quantities for biochemical and cellular assays.
  • Promotes actin-dependent dynamin oligomerization.
  • Reported HIV-1 integrase inhibitory activity.
  • Powder stable at -20°C for up to 3 years.
  • In-solution stability: -80°C for 6 months, -20°C for 1 month.
  • Available in small-mass pack sizes suitable for research assays.
  • Molecular formula C23H20N4O8; molecular weight 480.43 g/mol.