Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Tert-Butylcarbonyloxy)Iodobenzene

Catalog No. 66228325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-228-325G 25 g
66-228-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 66-228-325G Supplier Sigma Aldrich Supplier No. 66228325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(Tert-Butylcarbonyloxy)Iodobenzene

Specifications

CAS 57357-20-7
Melting Point 104°C to 109°C
Molecular Formula C16H23IO4
Linear Formula C16H23IO4
MDL Number MFCD08276835
Quantity 25 g
Synonym (Di-tert-butylcarbonyloxyiodo)benzol; 2,2-Dimethylpropanoic acid, phenyliodine complex; Bis(2,2-dimethylpropanoato-O)phenyliodide; Di-(Pivaloyloxy)iodobenzene
Molecular Weight (g/mol) 406.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.