Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tetrabutylammonium) Tetracyanodiphenoquinodimethanide 98.0+%, TCI America™
SDP

Catalog No. B1466100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1466100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1466100MG Supplier TCI America Supplier No. B1466100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68271-98-7
Molecular Formula C50H80N6
Molecular Weight (g/mol) 765.232
MDL Number MFCD00213933
InChI Key NJFQTNXYFOEFJU-UHFFFAOYSA-N
Synonym Bis(tetrabutylammonium) Tetracyanodiphenoquinodimethane Complex, TCNDQ
PubChem CID 44630139
IUPAC Name [2-[4-[4-(2-azanidylidene-1-cyanoethenyl)phenyl]phenyl]-2-cyanoethenylidene]azanide;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.C1=CC(=CC=C1C2=CC=C(C=C2)C(=C=[N-])C#N)C(=C=[N-])C#N

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 765.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tetrabutylammonium) Tetracyanodiphenoquinodimethanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.