Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis-(toluene-4-sulfonyl)methane

Catalog No. MNO0000071G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MNO0000071G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MNO0000071G Supplier Sigma Aldrich Supplier No. MNO0000071G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • In vitro activities of phenoxy and benzyloxyacetic acid derivatives as antisickling agents has been investigated by the application of quantitative structure activity relationship (qsar) of hansch-type.benzyloxyacetic acid may be used for the following syntheses:
  • Mixed benzyloxyacetic pivalic anhydride
  • Chiral glycolates, 1,2:4,5-di-o-isopropylidene-α-d-fructopyranos-3-yl benzyloxyacetate, via esterification
  • Tetraaqua(1,10-phenanthroline-[α]2n,nα)magnesium(ii) bis[(2,4-dichlorophenyl)acetate]
  • As ligand in the preparation of diaquabis(benzyloxyacetato)copper(ii) complexmay contain chloroacetic acid

Specifications

CAS 15310-28-8
Molecular Formula C15H16O4S2
Linear Formula C15H16O4S2
MDL Number MFCD00032255
Quantity 5 g
Molecular Weight (g/mol) 324.42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.