Learn More
Medchemexpress LLC Bis-Tos-PEG4 | 37860-51-8 | 502.60 | 100 G
SDP
Catalog No. 5000449088
Encompass_Preferred
Bis-Tos-PEG4 (PROTAC Linker 16) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • The other ligand binds to the target protein
  • For research use only
Catalog No. 50-004-49088 Supplier Medchemexpress LLC Supplier No. HYW004816100G
Bis-Tos-PEG4 (PROTAC Linker 16) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • The other ligand binds to the target protein
  • For research use only