Learn More
Medchemexpress LLC Bis-Tos-PEG4 (PROTAC linker 16) | 37860-51-8 | 502.60 | 500 G
SDP
Catalog No. 5000445507
Encompass_Preferred
Bis-Tos-PEG4 (PROTAC Linker 16) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • For research use only
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-45507 Supplier Medchemexpress LLC Supplier No. HYW004816500G
Bis-Tos-PEG4 (PROTAC Linker 16) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • For research use only
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins