Learn More
Medchemexpress LLC Bis-Tos-PEG7 | 69502-27-8 | 98.0% | C28H42O12S2 | 100 MG
SDP
Catalog No. 5000353144
Encompass_Preferred
Bis-Tos-PEG7 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53144 Supplier Medchemexpress LLC Supplier No. HY140368100MG
Bis-Tos-PEG7 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins