Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(tri-tert-butylphosphine)palladium(0), 98%

Catalog No. AC363500010
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363500010 1 g Glass bottle
AC363501000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC363500010 Supplier Thermo Scientific Chemicals Supplier No. 363500010
Only null left
Add to Cart

CAS: 53199-31-8 | C24H54P2Pd | 511.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53199-31-8
Molecular Formula C24H54P2Pd
Molecular Weight (g/mol) 511.06
MDL Number MFCD03094580
InChI Key MXQOYLRVSVOCQT-UHFFFAOYSA-N
Synonym bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
PubChem CID 2734558
SMILES [Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

Specifications

Melting Point 258.0°C to 272.0°C
Color Brown-Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pd)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
IUPAC Name palladium;tritert-butylphosphane
Formula Weight 511.05
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Bis(tri-tert-butylphosphine)palladium(0)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.