Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Trifluoromethane)Sulfonimide Lithium Salt

Catalog No. 44950450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-950-450G 50 g
44-950-410G 10 g
44-950-4250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 44-950-450G Supplier Sigma Aldrich Supplier No. 44950450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(Trifluoromethane)Sulfonimide Lithium Salt

Specifications

CAS 90076-65-6
Melting Point 234°C to 238°C
Molecular Formula C2F6LiNO4S2
Linear Formula CF3SO2NLiSO2CF3
MDL Number MFCD00210017
Quantity 50 g
Synonym Bis(trifluoromethylsulfonyl)amine lithium salt; Lithium bistrifluoromethanesulfonimidate
Molecular Weight (g/mol) 287.09 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.