Learn More
Medchemexpress LLC Bis(trimethylsilyl)acetylene | 14630-40-1 | MFCD00008276 | 99.0% | 170.40 g/mol | C8H18Si2 | 100 G
SDP
Catalog No. 5000301018
Encompass_Preferred
Bis(trimethylsilyl)acetylene is an organosilicon reagent used as a silyl-protected acetylene synthon in organic synthesis and research applications. It enables introduction of ethynyl fragments while the trimethylsilyl groups protect the alkyne intermediate during multistep sequences. The product is offered in laboratory pack sizes and is accompanied by quality documentation for research use.
  • Acts as a silyl-protected acetylene synthon for introducing ethynyl fragments.
  • Improves handling and stability compared with free acetylene.
  • Suitable for cross-coupling and cycloaddition reactions in synthetic chemistry.
  • Provided in multiple pack sizes to suit research needs.
  • High purity and accompanied by datasheet or certificate of analysis.
Catalog No. 50-003-01018 Supplier Medchemexpress LLC Supplier No. HYW016961100G
Bis(trimethylsilyl)acetylene is an organosilicon reagent used as a silyl-protected acetylene synthon in organic synthesis and research applications. It enables introduction of ethynyl fragments while the trimethylsilyl groups protect the alkyne intermediate during multistep sequences. The product is offered in laboratory pack sizes and is accompanied by quality documentation for research use.
  • Acts as a silyl-protected acetylene synthon for introducing ethynyl fragments.
  • Improves handling and stability compared with free acetylene.
  • Suitable for cross-coupling and cycloaddition reactions in synthetic chemistry.
  • Provided in multiple pack sizes to suit research needs.
  • High purity and accompanied by datasheet or certificate of analysis.