Learn More
Medchemexpress LLC Bis(trimethylsilyl)acetylene | 14630-40-1 | MFCD00008276 | 99.9% | 170.40 g/mol | C8H18Si2 | 10 G
SDP
Catalog No. 5000299879
Encompass_Preferred
Bis(trimethylsilyl)acetylene is an organosilicon reagent used in organic synthesis as a protected acetylene equivalent. It is commonly employed in research-scale coupling and alkynylation reactions to introduce alkyne functionality while minimizing handling of free acetylene.
  • Used as a masked acetylene for coupling and alkynylation reactions.
  • Facilitates introduction of alkyne functionality in complex molecule synthesis.
  • High purity: 99.9%.
  • Colorless to off-white; solid at lower temperatures and liquid at higher ambient temperatures.
  • Stable under recommended storage; long shelf life for the pure material.
Catalog No. 50-002-99879 Supplier Medchemexpress LLC Supplier No. HYW01696110G
Bis(trimethylsilyl)acetylene is an organosilicon reagent used in organic synthesis as a protected acetylene equivalent. It is commonly employed in research-scale coupling and alkynylation reactions to introduce alkyne functionality while minimizing handling of free acetylene.
  • Used as a masked acetylene for coupling and alkynylation reactions.
  • Facilitates introduction of alkyne functionality in complex molecule synthesis.
  • High purity: 99.9%.
  • Colorless to off-white; solid at lower temperatures and liquid at higher ambient temperatures.
  • Stable under recommended storage; long shelf life for the pure material.