Learn More
Medchemexpress LLC Bis(trimethylsilyl)acetylene | 14630-40-1 | MFCD00008276 | 99.9% | 170.40 g·mol⁻¹ | C8H18Si2 | 25 G
SDP
Catalog No. 5000303824
Encompass_Preferred
Bis(trimethylsilyl)acetylene is a silyl-protected acetylene reagent used as a masked alkyne source in organic synthesis and life-science research. It enables selective coupling and functionalization reactions while improving reagent stability and handling compared with free acetylene.
  • Acts as a masked acetylene for controlled alkyne incorporation.
  • High purity suitable for synthetic and analytical workflows.
  • Convenient physical handling and common laboratory pack sizes.
  • Compatible with common coupling and cycloaddition reactions.
  • Documentation available including COA and SDS for traceability.
Catalog No. 50-003-03824 Supplier Medchemexpress LLC Supplier No. 25GHYW016961
Bis(trimethylsilyl)acetylene is a silyl-protected acetylene reagent used as a masked alkyne source in organic synthesis and life-science research. It enables selective coupling and functionalization reactions while improving reagent stability and handling compared with free acetylene.
  • Acts as a masked acetylene for controlled alkyne incorporation.
  • High purity suitable for synthetic and analytical workflows.
  • Convenient physical handling and common laboratory pack sizes.
  • Compatible with common coupling and cycloaddition reactions.
  • Documentation available including COA and SDS for traceability.