Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis(trimethylsilyl)acetylene | 14630-40-1 | MFCD00008276 | 99.9% | 170.40 g·mol⁻¹ | C8H18Si2 | 5 G
SDP

Catalog No. 5000301230
Encompass_Preferred

Bis(trimethylsilyl)acetylene is an organosilicon reagent supplied for laboratory research and organic synthesis applications. It is provided in small lab pack sizes and characterized by confirmed purity, molecular data, and recommended cold storage for long-term stability.

  • Purity 99.87%.
  • Molecular formula C8H18Si2.
  • Molecular weight 170.40 g·mol⁻¹.
  • Appearance: solid below 21°C; liquid above 24°C.
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size available: 5 g.

Catalog No. 50-003-01230 Supplier Medchemexpress LLC Supplier No. HYW0169615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis(trimethylsilyl)acetylene is an organosilicon reagent supplied for laboratory research and organic synthesis applications. It is provided in small lab pack sizes and characterized by confirmed purity, molecular data, and recommended cold storage for long-term stability.

  • Purity 99.87%.
  • Molecular formula C8H18Si2.
  • Molecular weight 170.40 g·mol⁻¹.
  • Appearance: solid below 21°C; liquid above 24°C.
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size available: 5 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.