Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trimethylsilyl)methane, 98%

Catalog No. p-7022756
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1566206 5 g
AAA1566214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1566206 Supplier Thermo Scientific Chemicals Supplier No. A1566206
Only null left
Add to Cart

CAS: 2117-28-4 | C7H20Si2 | 160.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2117-28-4
Molecular Formula C7H20Si2
Molecular Weight (g/mol) 160.41
MDL Number MFCD00008275
InChI Key GYIODRUWWNNGPI-UHFFFAOYSA-N
Synonym bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl
PubChem CID 75029
IUPAC Name trimethyl(trimethylsilylmethyl)silane
SMILES C[Si](C)(C)C[Si](C)(C)C

Specifications

Boiling Point 132°C to 133°C
Melting Point -71°C
Quantity 5 g
UN Number UN1993
Beilstein 1734530
Flash Point 12°C (53°F)
Formula Weight 160.41
Percent Purity 98%
Refractive Index 1.417
Density 0.75
Chemical Name or Material Bis(trimethylsilyl)methane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 218-322-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.