Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trimethylsilyl)selenide 98%, Thermo Scientific™

Catalog No. AC291670050
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC291670050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC291670050 Supplier Thermo Scientific Chemicals Supplier No. 291670050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4099-46-1
Molecular Formula C6H18SeSi2
Molecular Weight (g/mol) 225.35
MDL Number MFCD00143781
InChI Key FKIZDWBGWFWWOV-UHFFFAOYSA-N
Synonym bis trimethylsilyl selenide, 1,1,1,3,3,3-hexamethyldisilaselenane, hexamethyldisilaselenane, bis-trimethylsilyl selenide, trimethyl trimethylsilylselanyl silane, 1,1,1,3,3,3-hexamethyldisilaselenane #, trimethyl trimethylsilyl selanyl silane
PubChem CID 553451
IUPAC Name trimethyl[(trimethylsilyl)selanyl]silane
SMILES C[Si](C)(C)[Se][Si](C)(C)C

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4800 to 1.4820 (20°C, 589nm)
Linear Formula (Si(CH3)3)2Se
Quantity 5 mL
Formula Weight 225.34
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl)selenide, 98%
missing translation for 'healthHazard1' MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.