Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(trimethylsilylmethyl) Sulfide 95.0+%, TCI America™
SDP

Catalog No. B46151ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46151ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B46151ML Supplier TCI America Supplier No. B46151ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4712-51-0
Molecular Formula C8H22SSi2
Molecular Weight (g/mol) 206.494
MDL Number MFCD00039797
InChI Key GISUCMFTNKRCPF-UHFFFAOYSA-N
PubChem CID 138328
IUPAC Name trimethyl(trimethylsilylmethylsulfanylmethyl)silane
SMILES C[Si](C)(C)CSC[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 85°C
Quantity 1 mL
Formula Weight 206.49
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilylmethyl) Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.