Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenylphosphine)copper(I) borohydride, Thermo Scientific™

Catalog No. AAA1373014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1373014 25 g
AAA1373022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1373014 Supplier Thermo Scientific Chemicals Supplier No. A1373014

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16903-61-0
Molecular Formula C36H34BCuP2
Molecular Weight (g/mol) 602.97
MDL Number MFCD00013219
InChI Key YVXRFTLDBYUBGJ-UHFFFAOYSA-N
Synonym bis triphenylphosphine copper tetrahydroborate, bis triphenylphosphine copper i borohydride, bis-triphenylphosphino-cuprous borohydride, tetrahydridoboratobis triphenylphosphine copper i, bis triphenylphosphine copper i tetrahydridoborate, bis triphenylphosphine copper tetrahydroborate,? pph3 2cubh4
PubChem CID 10908163
IUPAC Name boron(1-);copper(1+);triphenylphosphane
SMILES [BH4-].[Cu+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼175°C (decomposition)
Linear Formula [(C6H5)3P]2CuBH4
Quantity 25 g
UN Number UN2813
Sensitivity Moisture sensitive
Formula Weight 602.97
Chemical Name or Material Bis(triphenylphosphine)copper(I) borohydride
Hazard Category H261-H315-H319-H335
Hazard Statement GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
EINECSNumber 240-951-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.