Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenylphosphine)dicarbonylnickel, 98%

Catalog No. p-7050664
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3924227 2.5 g
AA3924209 10 g
AA3924218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3924227 Supplier Thermo Scientific Chemicals Supplier No. 03924227
Only null left
Add to Cart

CAS: 13007-90-4 | C38H30NiO2P2 | 639.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13007-90-4
Molecular Formula C38H30NiO2P2
Molecular Weight (g/mol) 639.30
MDL Number MFCD00009591
InChI Key SLFKPACCQUVAPG-UHFFFAOYSA-N
Synonym bis triphenylphosphine dicarbonylnickel
PubChem CID 22836333
IUPAC Name carbon monoxide;nickel;triphenylphosphanium
SMILES [Ni].[C-]#[O+].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 210°C to 215°C (decomposition)
Quantity 2.5 g
UN Number UN3466
Merck Index 14,1304
Formula Weight 639.29
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Bis(triphenylphosphine)dicarbonylnickel
Hazard Category H317-H350
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 235-851-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.