Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Triphenylphosphine)Palladium(II) Dichloride

Catalog No. 74124825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-124-825G 25 g
74-124-85G 5 g
74-124-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 74-124-825G Supplier Sigma Aldrich Supplier No. 74124825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(triphenylphosphine)palladium(II) dichloride is generally used as a catalyst for the synthesis of conjugated polymers via Stille coupling. It is also applicable to other coupling reactions such as Suzuki- and Sonogashira-coupling reactions

Specifications

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Linear Formula [(C6H5)3P]2PdCl2
MDL Number MFCD00009593
Quantity 25 g
Synonym Dichlorobis(triphenylphosphine)palladium(II); Palladium(II)bis(triphenylphosphine) dichloride; PdCl2(PPh3)2
Molecular Weight (g/mol) 701.9 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.