Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Triphenylphosphine)Palladium(II) Dichloride

Catalog No. 4127405G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
250 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-274-05G 5 g
41-274-01G 1 g
41-274-0250G 250 g
41-274-0250MG 250 mg
41-274-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Warning
Catalog No. 41-274-05G Supplier Sigma Aldrich Supplier No. 4127405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(triphenylphosphine)palladium(II) dichloride is an organometallic complex. It is an efficient cross-coupling catalyst for C-C coupling reaction, such as Negishi coupling, Suzuki coupling, Sonogashira coupling and Heck coupling reaction. Detection of bis(triphenylphosphine)palladium(II) dichloride by electrospray ionization quadrupole ion trap mass spectrometry using different imidazolium salts as the charge carrier has been reported. It is employed as catalyst for the Heck reaction medium.Bis(triphenylphosphine)palladium(II) dichloride was employed in the following studies:As model catalyst for the evaluation of functionalized silica'sas for the adsorptive recovery of homogeneous catalysts, via its interaction with metal centre.As catalyst in the synthesis of diphenylacetylene.One-pot synthesis of furoquinolines, via Pd-catalyzed Sonogashira coupling followed by Cu(I) catalyzed ring closure. Catalyst for Sonogashira coupling of aryl alkynes to 2-bromothiazole and 2-bromothiophene.

Specifications

CAS 13965-03-2
Molecular Formula C36H30Cl2P2Pd
Linear Formula [(C6H5)3P]2PdCl2
MDL Number MFCD00009593
Quantity 5 g
Synonym Dichlorobis(triphenylphosphine)palladium(II); Palladium(II)bis(triphenylphosphine) dichloride; PdCl2(PPh3)2
Molecular Weight (g/mol) 701.9 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.