Learn More
Gold Biotechnology Inc Bis Tris 100 g
SDP
Catalog No. 5000562062
Encompass_Preferred
Bis-Tris also known as bis-Tris methane is a zwitterionic buffering agent used in biochemistry and molecular biology It is a bis(2-hydroxyethyl)amine and Tris family buffer that is useful for a pH range of 5 8 7 2 Bis Tris is a common component of many buffer systems for various types of electrophoresis Bis-Tris may form a complex with some common metals such as Cu(II) and Pb(II) and formation constants should be taken into account when using this buffer in a solution containing metal ions Bis-Tris can be used as a substitute for the highly toxic buffer cacodylate
Catalog No. 50-005-62062 Supplier Gold Biotechnology Inc Supplier No. B020100
Bis-Tris also known as bis-Tris methane is a zwitterionic buffering agent used in biochemistry and molecular biology It is a bis(2-hydroxyethyl)amine and Tris family buffer that is useful for a pH range of 5 8 7 2 Bis Tris is a common component of many buffer systems for various types of electrophoresis Bis-Tris may form a complex with some common metals such as Cu(II) and Pb(II) and formation constants should be taken into account when using this buffer in a solution containing metal ions Bis-Tris can be used as a substitute for the highly toxic buffer cacodylate