Learn More
Gold Biotechnology Inc Bis-Tris 500 g
SDP
Catalog No. NC1539430
Encompass
Bis-Tris, also known as bis-Tris methane, is a zwitterionic buffering agent used in biochemistry and molecular biology. It is a bis(2-hydroxyethyl)amine and Tris family buffer that is useful for a pH range of 5.8 – 7.2. Bis Tris is a common component of many buffer systems for various types of electrophoresis. Bis-Tris may form a complex with some common metals, such as Cu(II) and Pb(II), and formation constants should be taken into account when using this buffer in a solution containing metal ions. Bis-Tris can be used as a substitute for the highly toxic buffer cacodylate.
Catalog No. NC1539430 Supplier Gold Biotechnology Inc Supplier No. B020500
Bis-Tris, also known as bis-Tris methane, is a zwitterionic buffering agent used in biochemistry and molecular biology. It is a bis(2-hydroxyethyl)amine and Tris family buffer that is useful for a pH range of 5.8 – 7.2. Bis Tris is a common component of many buffer systems for various types of electrophoresis. Bis-Tris may form a complex with some common metals, such as Cu(II) and Pb(II), and formation constants should be taken into account when using this buffer in a solution containing metal ions. Bis-Tris can be used as a substitute for the highly toxic buffer cacodylate.