Learn More
Bioworld Bis-Tris, 250 g
Supplier:  Bioworld 402200162
Encompass_Preferred
Synonym: 2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane A common biological bufferAn inert, zwitterionic buffer that exhibits only a very small shift in dissociation constantSpecifications:Ultra Pure GradepKa (25°C): 6.5+/-0.2Appearance: SolidBoiling Point : 466.18 °C at 760 mmHg (Predicted)Density : 1.35 g/cm3 (Predicted)Melting Point: 102-105 °CPH Value: 5.8-7.2
Catalog No. 50-255-447