Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis-Tris, cGMP Manufactured, 99%, Spectrum™ Chemical
SDP

Catalog No. 18601026 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-026 1 kg Poly Bottle
18-601-027 12 kg Poly Bottle
18-601-028 25 kg Fiber Drum
18-601-029 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-026 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B40921KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

B4092, 6976-37-0, C8H19NO5

Spectrum™ Chemical Bis-Tris, cGMP Manufactured - Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.24
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES OCCN(CCO)C(CO)(CO)CO

Specifications

CAS Max % 100%
Boiling Point >400°C / 752°F
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range 102-106°C
Packaging Poly Bottle
Quantity 1 kg
Percent Purity 99%
Grade cGMP, bioCERTIFIED
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Bis-Tris
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.