Learn More
Medchemexpress LLC Bis-tris-d19 | 352534-93-1 | 10 MG
SDP
Catalog No. 5000297785
Encompass_Preferred
Bis-Tris-d19 is the deuterium-labeled form of the Bis-Tris buffer, provided as a solid reagent intended for use as a tracer or internal standard in quantitative analysis and isotope-labeling studies. It is suitable for analytical applications such as NMR, GC-MS, and LC-MS and is supplied in a 10 mg vial.
  • Can be used as a tracer in research applications.
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • Deuterium labeling (d19) enables isotopic tracing and metabolic studies.
  • Supplied as a solid; recommended storage at -20°C for long-term stability.
Catalog No. 50-002-97785 Supplier Medchemexpress LLC Supplier No. HYD0856S10MG
Bis-Tris-d19 is the deuterium-labeled form of the Bis-Tris buffer, provided as a solid reagent intended for use as a tracer or internal standard in quantitative analysis and isotope-labeling studies. It is suitable for analytical applications such as NMR, GC-MS, and LC-MS and is supplied in a 10 mg vial.
  • Can be used as a tracer in research applications.
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • Deuterium labeling (d19) enables isotopic tracing and metabolic studies.
  • Supplied as a solid; recommended storage at -20°C for long-term stability.