Learn More
Medchemexpress LLC Bis-Tris-d19 | 352534-93-1 | 99% | 100 MG
SDP
Catalog No. 5000410599
Encompass_Preferred
Bis-Tris-d19 is the deuterium labeled Bis-Tris. Bis-Tris is a commonly used buffer. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-10599 Supplier Medchemexpress LLC Supplier No. HYD0856S100MG
Bis-Tris-d19 is the deuterium labeled Bis-Tris. Bis-Tris is a commonly used buffer. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.