Learn More
Medchemexpress LLC Bis-Tris hydrochlor 10g
SDP
Catalog No. 5000205022
Encompass_Preferred
Bis-Tris hydrochloride is a biological buffer and can be used in gel electrophoresis separations cell culture and bioprocessing applications[1]
Catalog No. 50-002-05022 Supplier Medchemexpress LLC Supplier No. HYW10568110G
Bis-Tris hydrochloride is a biological buffer and can be used in gel electrophoresis separations cell culture and bioprocessing applications[1]