Learn More
Medchemexpress LLC Bis-Tris hydrochlor 25g
SDP
Catalog No. 5000204865
Encompass_Preferred
Bis-Tris hydrochloride is a biological buffer and can be used in gel electrophoresis separations cell culture and bioprocessing applications[1]
Catalog No. 50-002-04865 Supplier Medchemexpress LLC Supplier No. 25GHYW105681
Bis-Tris hydrochloride is a biological buffer and can be used in gel electrophoresis separations cell culture and bioprocessing applications[1]