Learn More
Bioworld Bis-Tris Propane, 500 g
Catalog No. 50255450
Encompass_Preferred
A non-zwitterionic buffer that is useful in the pH 6.3-9.5 rangeSynonym: 1,3-Bis[tris(hydroxymethyl)methylamino]propane Appearance: SolidBoiling Point : 573.3 °CDensity : 1.326 g/cm3IUPAC_Name: 2,2'-(1,3-Propanediyldiimino)Bis[2-(Hydroxymethyl)-1,3-Propanediol]Melting Point: 164-165 °CPH Value: 6.3 - 9.5
Catalog No. 50-255-450 Supplier Bioworld Supplier No. 402200202
A non-zwitterionic buffer that is useful in the pH 6.3-9.5 rangeSynonym: 1,3-Bis[tris(hydroxymethyl)methylamino]propane Appearance: SolidBoiling Point : 573.3 °CDensity : 1.326 g/cm3IUPAC_Name: 2,2'-(1,3-Propanediyldiimino)Bis[2-(Hydroxymethyl)-1,3-Propanediol]Melting Point: 164-165 °CPH Value: 6.3 - 9.5