Learn More
AG Scientific Inc BIS-TRIS Propane 100 G
Catalog No. 502268248
Encompass_Preferred
BIS-TRIS Propane is a non-zwitterionic buffer that is useful in the pH 6.3-9.5 range.
Catalog No. 50-226-8248 Supplier AG Scientific Inc Supplier No. B2432100GM
BIS-TRIS Propane is a non-zwitterionic buffer that is useful in the pH 6.3-9.5 range.