Learn More
Medchemexpress LLC Bis-tris propane | 64431-96-5 | 99.9% | 282.33 | 10 G
SDP
Catalog No. 5000438140
Encompass_Preferred
Bis-tris propane (BTP) is a water-soluble buffer substance. It can serve as a suitable buffer for polymerase chain reaction (PCR) and has the ability to enhance the stability or activity of restriction enzymes.
  • Water-soluble buffer substance.
  • Suitable buffer for polymerase chain reaction (PCR).
  • Can enhance the stability or activity of restriction enzymes.
  • Demonstrates high fidelity in PCR reactions by stabilizing the pH of the reaction system and reducing errors introduced by DNA polymerase.
Catalog No. 50-004-38140 Supplier Medchemexpress LLC Supplier No. HY12639910G
Bis-tris propane (BTP) is a water-soluble buffer substance. It can serve as a suitable buffer for polymerase chain reaction (PCR) and has the ability to enhance the stability or activity of restriction enzymes.
  • Water-soluble buffer substance.
  • Suitable buffer for polymerase chain reaction (PCR).
  • Can enhance the stability or activity of restriction enzymes.
  • Demonstrates high fidelity in PCR reactions by stabilizing the pH of the reaction system and reducing errors introduced by DNA polymerase.