Learn More
Medchemexpress LLC Bis-tris propane | 64431-96-5 | 99.9% | 282.33 | 5 G
SDP
Catalog No. 5000438029
Encompass_Preferred
Bis-tris propane (BTP) is a water-soluble buffer substance that can be used as a suitable buffer for polymerase chain reaction (PCR) and can enhance the stability or activity of restriction enzymes. It exhibits high fidelity in PCR reactions by stabilizing the pH of the reaction system and reducing errors introduced by DNA polymerase.
  • Water-soluble buffer substance
  • Suitable for polymerase chain reaction (PCR)
  • Enhances stability or activity of restriction enzymes
  • Exhibits high fidelity in PCR reactions
  • Stabilizes pH of reaction system
  • Reduces errors introduced by DNA polymerase
Catalog No. 50-004-38029 Supplier Medchemexpress LLC Supplier No. HY1263995G
Bis-tris propane (BTP) is a water-soluble buffer substance that can be used as a suitable buffer for polymerase chain reaction (PCR) and can enhance the stability or activity of restriction enzymes. It exhibits high fidelity in PCR reactions by stabilizing the pH of the reaction system and reducing errors introduced by DNA polymerase.
  • Water-soluble buffer substance
  • Suitable for polymerase chain reaction (PCR)
  • Enhances stability or activity of restriction enzymes
  • Exhibits high fidelity in PCR reactions
  • Stabilizes pH of reaction system
  • Reduces errors introduced by DNA polymerase