Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BIS-Tris Propane, ULTROL™ Grade, Calbiochem™,

Catalog No. 394111100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-411-1100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-411-1100GM Supplier MilliporeSigma™ Supplier No. 394111100GM

missing translation for 'validMainframeMsg'

A non to zwitterionic buffer that is useful in the pH 6.3 to 9.5 range

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
Synonym 1,3-bis[tris(Hydroxymethyl)methylamino]propane
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO

Specifications

Quantity 100 g
Solution Type Buffer
Assay Percent Range >98%
Solubility Information Easily soluble in cold water
Formula Weight 282.4
Percent Purity ≥99% (titration)
Grade by titration
Chemical Name or Material non-zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.