Learn More
Sigma Aldrich Fine Chemicals Biosciences Bis24-di-tert-butylphenyl50MG
Catalog No. 502897479
Encompass
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at http//www.sigma aldrich.com
Catalog No. 50-289-7479 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9004850MG
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at http//www.sigma aldrich.com