Learn More
AdipoGen Bis4-bromophenyliodonium Tf
SDP
Catalog No. 502561635
Encompass_Preferred
Chemical. CAS 139139-81-4. Formula C13H8Br2F3IO3S. MW 587.97. Synthetic. Reagent for synthesis. Hypervalent iodine. Oxidizing agent.
Catalog No. 50-256-1635 Supplier AdipoGen Supplier No. CDXB0225G001
Chemical. CAS 139139-81-4. Formula C13H8Br2F3IO3S. MW 587.97. Synthetic. Reagent for synthesis. Hypervalent iodine. Oxidizing agent.