Learn More
AdipoGen Bis4methylphenyliodonium PF6
SDP
Catalog No. 502561624
Encompass_Preferred
Chemical. CAS 60565-88-0. Formula C14H14F6IP. MW 454.13. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-256-1624 Supplier AdipoGen Supplier No. CDXB0247G125
Chemical. CAS 60565-88-0. Formula C14H14F6IP. MW 454.13. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.