Learn More
Medchemexpress LLC HY-N4233 5mg Medchemexpress, Bisabolangelone CAS:30557-81-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN42335MG
Encompass_Preferred
Medchemexpress, HY-N4233 5mg Bisabolangelone CAS:30557-81-4 Bisabolangelone, a sesquiterpene derivative, is isolated from the roots of Osterici Radix. Bisabolangelone possesses anti-inflammatory properties, which inhibits LPS-stimulated inflammation through the blocking of NF-kappaB and MAPK pathways in macrophages. Bisabolangelone has anti-ulcer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2927