Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Bisabolol, ≥93% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019010 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9010 Supplier MilliporeSigma Supelco Supplier No. 954261ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(−)-α-Bisabolol is a monocyclic sesquiterpene alcohol naturally found in the essential oil of different plants like Vanillosmopsis erythropappa and Matricaria chamomilla. It finds applications in clinical and cosmetic industries due to its pharmacological properties like anti-inflammatory, anti-irritant, antimicrobial and skin healing property. It also shows effective action against malignant tumor cells. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 23089-26-1
Flash Point 135°C
Physical Form Neat
Linear Formula C15H26 O
MDL Number MFCD03412455
Synonym (-)-6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol; Levomenol
Type Analytical Standard
Quantity 1 mL
Formula Weight 222.37
Percent Purity ≥93% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.