Learn More
Medchemexpress LLC HY-B0557 500mg Medchemexpress, Bisacodyl CAS:603-50-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0557500MG
Encompass_Preferred
Medchemexpress, HY-B0557 500mg Bisacodyl CAS:603-50-9 Bisacodyl is a stimulant laxative agent that works directly on the colon to produce a bowel movement. Bisacodyl increases the secretion of PGE2 by direct activation of colon macrophages. PGE2 acts as a paracrine factor and decreases the expression of AQP3 in the colon, which inhibits water transfer from the luminal to the vascular side and leads to a laxative effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2598