Learn More
Apexbio Technology LLC Bisacodyl 603-50-9 5g
SDP
Catalog No. 502240549
Encompass_Preferred
Bisacodyl (CAS 603-50-9) is a synthetic diphenylmethane derivative classified as a stimulant laxative Its primary mechanism involves direct stimulation of enteric neurons within the colon leading to enhanced peristaltic contractions and increased colonic motility Bisacodyl also promotes accumulation of water and electrolytes in the intestinal lumen facilitating bowel evacuation In biomedical research bisacodyl is utilized to investigate gastrointestinal transit mechanisms and neuronal regulation of gut motility Its use supports studies focusing on intestinal physiology disorders of colonic motility and pharmacological modulation of the enteric nervous system
This item is not returnable. View return policy
Catalog No. 50-224-0549 Supplier Apexbio Technology LLC Supplier No. B18985000
Bisacodyl (CAS 603-50-9) is a synthetic diphenylmethane derivative classified as a stimulant laxative Its primary mechanism involves direct stimulation of enteric neurons within the colon leading to enhanced peristaltic contractions and increased colonic motility Bisacodyl also promotes accumulation of water and electrolytes in the intestinal lumen facilitating bowel evacuation In biomedical research bisacodyl is utilized to investigate gastrointestinal transit mechanisms and neuronal regulation of gut motility Its use supports studies focusing on intestinal physiology disorders of colonic motility and pharmacological modulation of the enteric nervous system