Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bisantrene dihydrochloride | 71439-68-4 | 98.9% | 471.39 g/mol | C22H24Cl2N8 | 25 MG
SDP

Catalog No. 5000211158
Encompass_Preferred

Bisantrene dihydrochloride is a research-grade antitumor compound that intercalates DNA and inhibits eukaryotic type II topoisomerases. Supplied as a solid for in vitro and preclinical research, it is used in cancer biology and pharmacology studies for mechanistic and efficacy assays.

  • High purity (98.85%) suitable for research use.
  • Confirmed molecular weight 471.39 g/mol and formula C22H24Cl2N8.
  • Available in multiple pack sizes, including 25 MG quantities.
  • Soluble in DMSO and water with recommended preparation procedures.
  • Store sealed and protect from moisture; observe solvent storage temperature guidelines.

Catalog No. 50-002-11158 Supplier Medchemexpress LLC Supplier No. HY100875A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bisantrene dihydrochloride is a research-grade antitumor compound that intercalates DNA and inhibits eukaryotic type II topoisomerases. Supplied as a solid for in vitro and preclinical research, it is used in cancer biology and pharmacology studies for mechanistic and efficacy assays.

  • High purity (98.85%) suitable for research use.
  • Confirmed molecular weight 471.39 g/mol and formula C22H24Cl2N8.
  • Available in multiple pack sizes, including 25 MG quantities.
  • Soluble in DMSO and water with recommended preparation procedures.
  • Store sealed and protect from moisture; observe solvent storage temperature guidelines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.