Learn More
Medchemexpress LLC Bisantrene dihydrochloride | 71439-68-4 | 98.9% | 471.39 g/mol | C22H24Cl2N8 | 5 MG
SDP
Catalog No. 5000211314
Encompass_Preferred
Bisantrene dihydrochloride is an anthracycline-class antitumor compound used in preclinical research. It intercalates into DNA, inhibits eukaryotic type II topoisomerases, and has demonstrated cytotoxic activity across multiple experimental tumor models. The material is provided as a solid for laboratory use with manufacturer storage and solubility guidance for solution preparations.
Catalog No. 50-002-11314 Supplier Medchemexpress LLC Supplier No. HY100875A5MG
Bisantrene dihydrochloride is an anthracycline-class antitumor compound used in preclinical research. It intercalates into DNA, inhibits eukaryotic type II topoisomerases, and has demonstrated cytotoxic activity across multiple experimental tumor models. The material is provided as a solid for laboratory use with manufacturer storage and solubility guidance for solution preparations.