Learn More
Medchemexpress LLC Bisantrene dihydrochloride | 71439-68-4 | MFCD01743164 | >98.0% | 471.39 g/mol | C22H24Cl2N8 | 100 MG
SDP
Catalog No. 5000211496
Encompass_Preferred
Bisantrene dihydrochloride is the dihydrochloride salt of bisantrene, an anthracene-derived antitumor agent used in research for its DNA intercalation and topoisomerase II inhibitory activity. It is supplied as a solid for laboratory use and is suitable for in vitro and preclinical studies where a high-purity research reagent is required.
  • Dihydrochloride salt form improves aqueous solubility for experimental work.
  • High purity (≥98% HPLC) suitable for biological and analytical assays.
  • Molecular weight 471.39 g/mol supports accurate dosing and calculations.
  • Acts as a DNA intercalator and topoisomerase II inhibitor for mechanistic studies.
  • Provided as a solid convenient for handling, storage, and formulation.
Catalog No. 50-002-11496 Supplier Medchemexpress LLC Supplier No. HY100875A100MG
Bisantrene dihydrochloride is the dihydrochloride salt of bisantrene, an anthracene-derived antitumor agent used in research for its DNA intercalation and topoisomerase II inhibitory activity. It is supplied as a solid for laboratory use and is suitable for in vitro and preclinical studies where a high-purity research reagent is required.
  • Dihydrochloride salt form improves aqueous solubility for experimental work.
  • High purity (≥98% HPLC) suitable for biological and analytical assays.
  • Molecular weight 471.39 g/mol supports accurate dosing and calculations.
  • Acts as a DNA intercalator and topoisomerase II inhibitor for mechanistic studies.
  • Provided as a solid convenient for handling, storage, and formulation.