Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bisantrene dihydrochloride | 71439-68-4 | MFCD01743164 | >98.0% | 471.39 g/mol | C22H24Cl2N8 | 50 MG
SDP

Catalog No. 5000211759
Encompass_Preferred

Bisantrene dihydrochloride is the dihydrochloride salt of bisantrene, an anthracycline-class antitumor agent that intercalates DNA and inhibits topoisomerase. It is supplied as a research-grade solid for in vitro and cellular studies of DNA damage and cell-cycle pathways, and requires sealed, moisture-free storage with low-temperature solvent storage for extended stability.

  • Anthracycline-class mechanism: DNA intercalation and topoisomerase inhibition.
  • Research applications: cellular assays, DNA damage, and cell-cycle studies.
  • High purity suitable for analytical and biological assays.
  • Solid form supplied in sealed quantities for laboratory use.
  • Storage guidance: sealed at 4°C short term; in solvent -20°C (1 month) and -80°C (up to 6 months).
  • Molecular formula C22H24Cl2N8; molecular weight 471.39 g/mol.

Catalog No. 50-002-11759 Supplier Medchemexpress LLC Supplier No. HY100875A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bisantrene dihydrochloride is the dihydrochloride salt of bisantrene, an anthracycline-class antitumor agent that intercalates DNA and inhibits topoisomerase. It is supplied as a research-grade solid for in vitro and cellular studies of DNA damage and cell-cycle pathways, and requires sealed, moisture-free storage with low-temperature solvent storage for extended stability.

  • Anthracycline-class mechanism: DNA intercalation and topoisomerase inhibition.
  • Research applications: cellular assays, DNA damage, and cell-cycle studies.
  • High purity suitable for analytical and biological assays.
  • Solid form supplied in sealed quantities for laboratory use.
  • Storage guidance: sealed at 4°C short term; in solvent -20°C (1 month) and -80°C (up to 6 months).
  • Molecular formula C22H24Cl2N8; molecular weight 471.39 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.