Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BisBenzimide H 33258, Sigma-Aldrich

Catalog No. 01258925 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-925 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-925 Supplier MilliporeSigma Supelco Supplier No. B2883500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

bisBenzimide H 33258 is a fluorescent stain for DNA, chromosomes and nuclei.

  • Suitable for fluorescence microscopy and flow cytometry.
  • Used for nuclear staining in cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23491-45-4

Specifications

Quantity 500 mg
Product Type Stains, bioactive small molecules
Purity ≥98%
Label Type Other Labels or Dyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.