Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BisBenzimide H 33342 trihydrochloride, Sigma-Aldrich

Catalog No. 01258855 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-855 1 g
01-258-995 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-855 Supplier MilliporeSigma Supelco Supplier No. B22611G
Add to Cart
Add to Cart

Fluorescent properties of bisBenzimide H 33342:

  • Free dye: Excitation maximum = 340 nm
  • Emission maximum = 510 nm (5 mM HEPES, 10 mM NaCl, pH 7. 0)
  • DNA complex: Excitation maximum = 355 nm
  • Emission maximum = 465 nm (5 mM HEPES, 10 mM NaCl, pH 7. 0)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875756-97-1

Specifications

Quantity 1 g
Product Type Fluorescence Reagent
Molecular Weight (g/mol) 561.93 g/mol, 561.94
Purity ≥98%
Molecular Formula C27H31Cl3N6O
InChI Key JABNPSKWVNCGMX-UHFFFAOYSA-N
SMILES Cl.Cl.Cl.CCOC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NC2=CC=C(C=C2N1)N1CCN(C)CC1
IUPAC Name 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-1,3-benzodiazol-2-yl]-1H-1,3-benzodiazole trihydrochloride
Label Type Other Labels or Dyes
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.