Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bisbenzodioxolo[5,6-a:5′,6′-g]quinolizinium, 5,6-dihydro-, chloride | 30044-78-1 | 25 MG
SDP

Catalog No. 5000420332
Encompass_Preferred

Pseudocoptisine chloride, also known as Isocoptisine chloride, is a quaternary alkaloid derived from Corydalis Tuber. This compound acts as an acetylcholinesterase (AChE) inhibitor and demonstrates anti-inflammatory and anti-amnestic properties. It is supplied as a yellow to brown solid with a purity of 99.36%.

  • Inhibits acetylcholinesterase (AChE) activity with an IC50 of 12.8 μM
  • Exhibits anti-inflammatory effects
  • Exhibits anti-amnestic effects
  • Molecular weight: 355.77
  • Chemical formula: C19H14ClNO4
  • Inhibits cell proliferation in KM12C cells
  • Reduces LPS-induced nitric oxide production in RAW264.7 cells
  • Suppresses TNF-α and IL-6 production

Catalog No. 50-004-20332 Supplier Medchemexpress LLC Supplier No. 25MGHYN6894A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pseudocoptisine chloride, also known as Isocoptisine chloride, is a quaternary alkaloid derived from Corydalis Tuber. This compound acts as an acetylcholinesterase (AChE) inhibitor and demonstrates anti-inflammatory and anti-amnestic properties. It is supplied as a yellow to brown solid with a purity of 99.36%.

  • Inhibits acetylcholinesterase (AChE) activity with an IC50 of 12.8 μM
  • Exhibits anti-inflammatory effects
  • Exhibits anti-amnestic effects
  • Molecular weight: 355.77
  • Chemical formula: C19H14ClNO4
  • Inhibits cell proliferation in KM12C cells
  • Reduces LPS-induced nitric oxide production in RAW264.7 cells
  • Suppresses TNF-α and IL-6 production

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.