Learn More
Cayman Chemical Bisdemethoxycurcumin, Purity: ≥98%, CAS: 33171-05-0, 50 mg
SDP
Catalog No. 502951171
Encompass_Preferred
A natural demethoxy derivative of curcumin; activates macrophage phagocytosis and, with 1α,25-dihydroxy vitamin D3, stimulates amyloid-β clearance by macrophages (optimal stimulation at 100 nM BDMC); suppresses proliferation in cancer cells; down-regulates p300 and inhibits LPS-induced iNOS expression
Catalog No. 50-295-1171 Supplier Cayman Chemical Supplier No. 1096050 MG
A natural demethoxy derivative of curcumin; activates macrophage phagocytosis and, with 1α,25-dihydroxy vitamin D3, stimulates amyloid-β clearance by macrophages (optimal stimulation at 100 nM BDMC); suppresses proliferation in cancer cells; down-regulates p300 and inhibits LPS-induced iNOS expression